Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Mouse AGRP Guinea Pig Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

GP-029-50
BSENGP-029-50EA 312 EUR InStock
BSENGP-029-50
Anti-Mouse AGRP Guinea Pig Polyclonal Antibody
Antikörper
AGRP is the endogenous antagonist of alpha-melanocyte stimulating hormone and has been shown to cause potent stimulation of food intake, and this protein is found in over 90% of Neuropeptide Y containing cells in rats.

Specificity was demonstrated by immunohistochemistry.
Application Information:
IHC, frozen, PFA fixed material. Not yet tested on paraffin embedded tissue sections. A concentration of 1:1000 to 1:2000 is recommended for IHC with overnight incubations. Permeabilization suggested is 0.1% triton X-100 in blocking buffer. IHC performed in sheep brain (hypothalamus) demonstrates intense staining of cells and terminals. No staining is evident when the primary antibody is pre-absorbed with 0.5 mg/ml of AGRP. In rodents such as rat cell terminal staining is most typically observed without cell body staining. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Mouse AGRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: carboxy domain of Mouse agouti related protein
Host: Guinea Pig
Isotype: IgG
Reactivity: Human, Rat, Sheep

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge