Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FBL Mouse Monoclonal Antibody [clone: 38F3]

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

M-1372-250
BSENM-1372-250EA 312 EUR InStock
BSENM-1372-250
Anti-FBL Mouse Monoclonal Antibody [clone: 38F3]
Antikörper
Fibrillarin is part of the small subunit processome complex, involved in the processing of pre-18S ribosomal RNA. Nop1p is the yeast homologue of fibrillarin. Fibrillarin/Nop1p is extraordinarily conserved, so that the yeast and human proteins are 67% identical, and the human protein can functionally replace the yeast protein. This means that suitably cross-reactive antibodies to Nop1p/fibrillarin such as this antibody can be used to reveal nucleoli and study fibrillarin/Nop1p in all eukaryotes and archea tested to date. This antibody is becoming widely used as a convenient marker for nucleoli in a wide variety of species.

Western Blotting (WB), Immunocytochemistry (IC). A dilution of 1:2000 (cell lysates) to 1:10000 (nuclear fractions) is recommended for WB of yeast protein samples followed by chemiluminescent detection (ECL). For other non-ECL WB methods, a dilution of 1:1000 to 1:5000 is recommended. A dilution of 1:500 is recommended for ECL WB on mammalian fibrillarin. A dilution of 1:1000 to 1:5000 is recommended for IC on yeast cells and 1:500 for mammalian cells. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FBL
Clonality: Monoclonal
Clone: 38F3
Conjugation: Unconjugated
Epitope: Yeast nuclear preparations. Hybridomas were screened by immunofluorescence on yeast cells and by western blotting on yeast protein homogenates (S. cerevisiae).
Host: Mouse
Isotype:
Reactivity: Human;Rat;Drosophila;S. pombe;C. elegans;S. pombe

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge