Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Nestin Mouse Monoclonal Antibody [clone: 4D11]

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

M-1385-100
BSENM-1385-100EA 312 EUR InStock
BSENM-1385-100
Anti-Nestin Mouse Monoclonal Antibody [clone: 4D11]
Antikörper
Nestin is a member of the class IV intermediate filament protein family which is expressed in neuronal stem cells. The molecular weight of human Nestin as determined by SDS-PAGE mobility is about 240kDa. However the real molecular weight is considerably less than this, at 177kDa, the disparity being likely due to the highly charged region of the C-terminal segment. Nestin is relatively poorly conserved in protein sequence across species boundaries, so that the mouse and human proteins have an overall identity of only 62%. As a result antibodies to the human protein often fail to recognize the rodent homologue and vice versa. However this antibody stains both rodent and human Nestin. Antibodies to Nestin are widely used to identify neural stem cells.

This antibody is specific for the 240 kDa Nestin protein by WB on developing rat brain (P18) homogenate. A much weaker band at approx. 90 kDa may also be seen. This is suggested to be a breakdown product of the 240 kDa band.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). Suggested dilution for WB is 1:1,000-5,000 and 1:250-500 for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Nestin
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rodent

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge