Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-14-3-3 Mouse Monoclonal Antibody [clone: 3G12]

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

M-1572-100
BSENM-1572-100EA 312 EUR InStock
BSENM-1572-100
Anti-14-3-3 Mouse Monoclonal Antibody [clone: 3G12]
Antikörper
14.3.3 protein eta or 14.3.3  binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner (Ref SwissProt). 14.3.3 protein eta is widely expressed as both homodimers and heterodimers and are concentrated in the nervous system. High concentrations of 14.3.3 protein eta have been linked to Creutzfeld Jacob Disease, Parkinson's Disease and early-onset schizopherenia.

Antibody recognizes 14.3.3 protein by western blot, immunocytochemistry on acetone or 4% PFA fixed cells, or frozen tissues, not yet tested on paraffin fixed tissues. Approved applicaitons include: WB, IHC/IF. Suggested dilution of 1:500-1:1,000 for IHC/IF. Suggested dilution of 1:1,000-1:5,000 for WB. A suitable control tissue is rat spinal cord or peripheral nerve homogenate.
Application Information:
WB, IHC/IF. Suggested dilution of 1:500-1:1,000 for IHC/IF. Suggested dilution of 1:1,000-1:5,000 for WB. A suitable control tissue is rat spinal cord or peripheral nerve homogenate.

Type: Primary
Antigen: 14-3-3
Clonality: Monoclonal
Clone: 3G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge