Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ChAT Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-043-100
BSENR-043-100EA 302 EUR InStock
BSENR-043-100
Anti-ChAT Rabbit Polyclonal Antibody
Antikörper
This gene encodes an enzyme which catalyzes the biosynthesis of the neurotransmitter acetylcholine. This gene product is a characteristic feature of cholinergic neurons, and changes in these neurons may explain some of the symptoms of Alzheimer's disease. Polymorphisms in this gene have been associated with Alzheimer's disease and mild cognitive impairment. Mutations in this gene are associated with congenital myasthenic syndrome associated with episodic apnea. Multiple transcript variants encoding different isoforms have been found for this gene, and some of these variants have been shown to encode more than one isoform. [provided by RefSeq, May 2010]

This antiserum stains cholinergic neurons in guinea-pig and rabbit.
Application Information:
Immunohistochemistry/cytochemistry on 4% PFA or formalin fixed frozen sections; paraffin sections can be more difficult and require more extensive antigen recovery methods. 1:400-1:2000 depending up detection and incubation times
ELISA: direct antigen ELISA
This antiserum will superbly stain both cell bodies and nerve terminal, and works particularly well in enteric and peripheral neurons. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ChAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: porcine ChAT protein (aa: 167-188)
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig, Mouse, Rat, Rabbit

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge