Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-native DBH Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-108-100
BSENR-108-100EA 302 EUR InStock
BSENR-108-100
Anti-native DBH Rabbit Polyclonal Antibody
Antikörper
DBH is an oxireductase belonging to the copper type II ascorbate-dependent monooxygenase family. DBH exists as a homotetramer composed of two non-covalently bound disulfide-linked dimers. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It binds 2 copper ions and 1 PQQ per subunit . Depending on the presence of a signal peptide, DBH can exist in both soluble and membrane-bound forms.

This antiserum has been extensively characterised in several laboratories. It reacts with a single band on western blot of bovine adrenal homogenates and stains only cells known to contain DBH, such as sympathetic neurons, adrenal medullary cells and central adrenergic neurons. Staining is abolished by preincubation with the enzyme.
Application Information:
IHC (frozen), Western Blot, radioimmunoassay and inhibition of enzymatic activity. Recommended to be used at a dilution of 1:500 to 1: 2000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: native DBH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Guinea Pig, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge