Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SUMO-1 Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-115-100
BSENR-115-100EA 312 EUR InStock
BSENR-115-100
Anti-SUMO-1 Rabbit Polyclonal Antibody
Antikörper
FUNCTION: Ubiquitin-like protein which binds to a wide range of target proteins. Does not seem to be involved in protein degradation and may function as an antagonist of ubiquitin in the degradation process. Plays a role in a number of cellular processes such as nuclear transport, DNA replication and repair, mitosis and signal transduction. Involved in targeting RANGAP1 to the nuclear pore complex protein RANBP2. SUBUNIT: Covalently attached to a number of proteins such as PML, RANGAP1, HIPK2, SP100, p53, p73alpha, MDM2, JUN and DNMT3B. Also interacts with HIF1A, HIPK2, HIPK3, CHD3, PIAS1, EXOSC9, TDG, RAD51 and RAD52. SUBCELLULAR LOCATION: Nucleus; nuclear membrane. Nucleus; nucleoplasm; nuclear speckle. Cytoplasm. SIMILARITY: Belongs to the ubiquitin family. SMT3 subfamily. SIMILARITY: Contains 1 ubiquitin-like domain.

This antiserum recognises human SUMO-1 and not ubiquitin.
Application Information:
IHC, WB. A dilution of 1:1000 to 1:2000 is recommended for immunohistochemistry and 1:2000 to 1:4000 for western blot. Cell lysate from Hela and NIH-3T3 cell lysates may be used as a positive control, and for IHC, lung carcinoma may be used. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SUMO-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa: 6-21 Human SUMO-1
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge