Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Synphilin-1 Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-117-100
BSENR-117-100EA 302 EUR InStock
BSENR-117-100
Anti-Synphilin-1 Rabbit Polyclonal Antibody
Antikörper
Synuclein alpha interacting protein (Synphilin-1) contains several protein-protein interaction domains and interacts with alpha synuclein in neurons. Mutations of SNCAIP have been linked to Parkinson disease. The amino acid sequence of synphilin-1 shares a high level of identity with its human counterpart, particularly in regions containing ankyrin-like motifs and the coiled-coil domain. Expression pattern of synphilin-1 in tissues is similar in both mouse and human. Synphilin-1 has an important role in the formation of aggregates and cytotoxicity in Parkinson disease and also Dorfin may be involved in the pathogenic process by ubiquitylation of synphilin-1.

A high level of specificity has been shown for this antiserum by IHC on human brain.
Application Information:
IHC, WB. A dilution of 1: 1000 to 1:2000 is recommended for immunohistochemistry and 1:500-1:1000 for western blot. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Synphilin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge