Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Tyrosine Kinase Receptor B Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-121-100
BSENR-121-100EA 302 EUR InStock
BSENR-121-100
Anti-Tyrosine Kinase Receptor B Rabbit Polyclonal Antibody
Antikörper
FUNCTION: Receptor for brain-derived neurotrophic factor (BDNF), neurotrophin-3 and neurotrophin-4/5 but not nerve growth factor (NGF). Involved in the development and/or maintenance of the nervous system. This is a tyrosine-protein kinase receptor. Known substrates for the TRK receptors are SHC1, PI-3 kinase, and PLC-gamma-1. CATALYTIC ACTIVITY: ATP + a [protein]-L-tyrosine = ADP + a [protein]-L-tyrosine phosphate. SUBUNIT: Exists in a dynamic equilibrium between monomeric (low affinity) and dimeric (high affinity) structures (By similarity). Binds APS. Interacts with SQSTM1. SUBCELLULAR LOCATION: Membrane; single-pass type I membrane protein. ALTERNATIVE PRODUCTS: 3 named isoforms produced by alternative splicing. Additional isoforms seem to exist. TISSUE SPECIFICITY: The different forms are differentially expressed in various cell types. Isoform T2 is primarily expressed in neurons. PTM: Ligand-mediated auto-phosphorylation. SIMILARITY: Belongs to the Tyr protein kinase family. Insulin receptor subfamily. SIMILARITY: Contains 2 Ig-like C2-type (immunoglobulin-like) domains. SIMILARITY: Contains 2 LRR (leucine-rich) repeats. SIMILARITY: Contains 1 protein kinase domain.

Sequence was chosen to avoid cross reactivity with TrkA and TrkC.
Application Information:
IHC. A dilution of 1:1000 to 1:3000 is recommended for this application. The sequence of the immunogen and the conjugate are identical to the one used by Yan et al (see the Refs) and the antiserum appears to have similar characteristics. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Tyrosine Kinase Receptor B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: rat mature TrkB protein (aa: 23-36)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge