Anti-Maltose Binding Protein Tag Rabbit Polyclonal Antibody
Lieferant: Biosensis
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R-1313-100
BSENR-1313-100EA 388 EUR InStock
BSENR-1313-100
Anti-Maltose Binding Protein Tag Rabbit Polyclonal Antibody
Antikörper
Maltose binding protein (MBP) is encoded by the malE gene of E.coli. MBP is often used in protein expression studies because it creates a stable fusion product that does not appear to interfere with the bioactivity of the protein of interest. It also allows for its easy purification from bacterial extracts under mild conditions.
This antibody recognizes the MBP epitope tag fused to the amino- or carboxy- termini of targeted proteins.
Application Information:
Western Blotting (WB) and ELISA. Suggested starting dilutions for WB of 1:1,000. Biosensis recommends optimal dilutions/More Product Information
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..