Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GABARAP Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-143-100
BSENR-143-100EA 302 EUR InStock
BSENR-143-100
Anti-GABARAP Rabbit Polyclonal Antibody
Antikörper
GABARAP is highly positively charged in its N-terminus and shares sequence homology with MAP1LC3 1A and 1B. This protein clusters neurotransmitter receptors (GABA(A) receptors) by mediating interaction with the cytoskeleton.  SUBUNIT: Interacts with GABRG2, TUBA1, ULK1 and NSF. Interacts with beta-tubulin and GPHN. SUBCELLULAR LOCATION: Intracytoplasmic membrane. Cytoskeleton. Largely associated with intracellular membrane structures including the Golgi apparatus and post-synaptic cisternae. Colocalizes with microtubules. TISSUE SPECIFICITY: Heart, brain, placenta, skeletal muscle, kidney and pancreas.

IHC, WB and ELISA confirmed the specificity for GABARAP.
Application Information:
IHC, immunofluorescence, WB. A dilution of 1:200 to 1:1000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GABARAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal of Human GABARAP
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge