Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Somatostatin Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-1502-50
BSENR-1502-50EA 321 EUR InStock
BSENR-1502-50
Anti-Somatostatin Rabbit Polyclonal Antibody
Antikörper
Somatostatin is secreted by neuroendocrine neurons in hypothalamus and interacts with somatostatin receptors inhibiting secretion of somatotropin.

Antibody recognizes Somatostatin in native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross reactivity: highly conserved, human, mouse and rat tested to date, other species are expected because of sequence similarity.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Somatostatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Somatostatin peptide (103-116)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge