Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-the Tyrosine Kinase Receptor C ECD Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-151-100
BSENR-151-100EA 302 EUR InStock
BSENR-151-100
Anti-the Tyrosine Kinase Receptor C ECD Rabbit Polyclonal Antibody
Antikörper
TrkC is a member of the neurotrophic tyrosine receptor kinase family. TrkC is a membrane-bound receptor that upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. TrkC is the receptor for neurotrophin-3 (NT-3).  Signalling through TrkC leads to cell differentiation and may play a role in the development of proprioceptive neurons that sense body position. SUBUNIT: Exists in a dynamic equilibrium between monomeric (low affinity) and dimeric (high affinity) structures. SUBCELLULAR LOCATION: Membrane; single-pass type I membrane protein. ALTERNATIVE PRODUCTS: 4 named isoforms produced by alternative splicing. Additional isoforms seem to exist. Mutations in TrkC have been associated with medulloblastomas, secretory breast carcinomas and other cancers.

Specificity was demonstrated by immunohistochemistry. This antibody was used to stain cryostat sections of the rat peripheral sensory ganglia.
Application Information:
IHC. A dilution of 1:1000 to 1:3000 is recommended for this application. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: the Tyrosine Kinase Receptor C ECD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge