Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Leptin Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-1553-100
BSENR-1553-100EA 312 EUR InStock
BSENR-1553-100
Anti-Leptin Rabbit Polyclonal Antibody
Antikörper
Leptin is secreted by white adipocytes and functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. Leptin has a variety of other roles including endocrine functions, regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing (Ref Entrez Gene).

Antibody recognizes rodent leptin with low cross reactivity to human leptin in WB and ELISA. Suggested dilution of 1:1,000-1:4,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Application Information:
WB and ELISA. Suggested dilution of 1:1,000-1:4,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge