Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-rh NTN Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-1561-100
BSENR-1561-100EA 312 EUR InStock
BSENR-1561-100
Anti-rh NTN Rabbit Polyclonal Antibody
Antikörper
Neurturin (NTN) is a member of the GDNF family of neurotrophic factors. This protein is a potent survival factor for several populations of central and peripheral neurons in mature and developing rodents. FUNCTION: Supports the survival of sympathetic neurons in culture. May regulate the development and maintenance of the CNS. Might control the size of non-neuronal cell population such as haemopoietic cells. SUBUNIT: Homodimer; disulfide-linked. SUBCELLULAR LOCATION: Secreted protein. DISEASE: Defects in NRTN are a cause of Hirschsprung disease (HSCR). In association with mutations of RET gene, and possibly with other loci, defects in NRTN are involved in Hirschsprung disease. This genetic disorder of neural crest development is characterized by the absence of intramural ganglion cells in the hindgut, often resulting in intestinal obstruction. SIMILARITY: Belongs to the TGF-beta family. GDNF subfamily.

Specificity of this product was demonstrated by immunohistochemistry and dot blot showed no cross reactivity with GDNF.
Application Information:
Immunohistochemistry (IHC) at a suggested dilution of 1:2000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: rh NTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge