Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Peroxiredoxin-3 Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-1592-100
BSENR-1592-100EA 312 EUR InStock
BSENR-1592-100
Anti-Peroxiredoxin-3 Rabbit Polyclonal Antibody
Antikörper
Peroxiredoxin-3 has a role in redox regulation of the cell.

Antibody recognizes Perioxiredoxin-3 in multiple applications including IHC, WB, ELISA. This antibody works in IHC on frozen or wax embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for light microscopy are 1:500 to 1:1,000 and IF of 1:50 to 1:100. For WB a dilution range of 1: 1,000 to 1: 2,000 is recommended. This antibody detects a protein at approx 23kDa on WB of human brain tissue which is the mature form of this protein. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
IHC, WB, ELISA. This antibody works in IHC on frozen or wax embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for light microscopy are 1:500 to 1:1,000 and IF of 1:50 to 1:100. For WB a dilution range of 1: 1,000 to 1: 2,000 is recommended. This antibody detects a protein at approx 23kDa on WB of human brain tissue which is the mature form of this protein. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Peroxiredoxin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: (127-143 aa) on Human Peroxiredoxin-3
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat,

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge