Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Glyceraldehyde 3-Phosphate Dehydrogenase/GAPDH Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-1701-100
BSENR-1701-100EA 302 EUR InStock
BSENR-1701-100
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase/GAPDH Rabbit Polyclonal Antibody
Antikörper
Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) is a metabolic enzyme responsible for catalyzing one step in the glycolytic pathway, the reversible oxidative phosphorylation of glyceraldehyde 3-phosphate. GAPDH may have other roles in the activation of transcription and in the regulation of apoptosis as well as Alzheimer's disease and Huntington's disease. The immunogen used to raise this particular antibody was extensively purified pig GAPDH. This antibody can be used as a loading control for western blotting experiments, allowing comparison between the level of this protein and others in a cell or tissue.

Specific for the ~36-38kDa Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) protein.
Application Information:
Western Blotting (WB) and Immunocytochemistry (ICC). A dilution of 1:10,000-1:30,000 is recommended for WB. Human GAPDH has a predicted length of 335 residues and a MW of 36 kDa. A dilution of 1:1,000-1:10,000 is recommended for ICC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase/GAPDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge