Anti-RBFOX3 Rabbit Polyclonal Antibody
Lieferant: Biosensis
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R-3771-100
BSENR-3771-100EA 312 EUR InStock
BSENR-3771-100
Anti-RBFOX3 Rabbit Polyclonal Antibody
Antikörper
Rabbit anti-neuronal nuclei antigen/Fox3 (NeuN/Fox3) polyclonal antibody (unconjugated), suitable for WB, IHC, ICC.
  • Human, mouse, rat
Applications: Western Blotting (WB), Immunocytochemistry (ICC) and Immunohistochemistry (IHC). A dilution of 1:500 - 1:1,000 is recommended for WB. A dilution of 1:5,000 - 1:10,000 is recommended for ICC and IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge