Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-beta Amyloid Rabbit Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R-959-100
BSENR-959-100EA 302 EUR InStock
BSENR-959-100
Anti-beta Amyloid Rabbit Polyclonal Antibody
Antikörper
The beta Amyloid peptide is derived from the cleavage of the Amyloid precursor protein and varies in length from 39 to 43 amino acids. Beta amyloid peptides are the major constituents of the plaques and tangles that occur in Alzheimer's disease.

Antibody recognizes human beta amyloid peptide in ELISA, other applications and other species not yet tested.
Application Information:
ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: beta Amyloid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acid sequence 1-14 of mature Human beta amyloid
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge