Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-rh Basic FGF Sheep Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

S-008-100
BSENS-008-100EA 302 EUR InStock
BSENS-008-100
Anti-rh Basic FGF Sheep Polyclonal Antibody
Antikörper
Fibroblast growth factors (FGFs), a heparin binding growth factor, exhibit widespread mitogenic and neurotrophic activities in a variety of different cells including mesenchymal, neuroectodermal and endothelial cells. aFGF (FGF-1) and bFGF (FGF-2) are present in relatively high levels in CNS. aFGF is expressed by a subset of neuronal populations, while bFGF is expressed by astrocytes, both lack signal peptides. Human bFGF is a 17.2 kDa protein containing 155 amino acid residues. FUNCTION: The heparin-binding growth factors are angiogenic agents in vivo and are potent mitogens for a variety of cell types in vitro. There are differences in the tissue distribution and concentration of these 2 growth factors. SUBUNIT: Monomer. Interacts with CSPG4 and FGFBP1. Found in a complex with FGFBP1, FGF1 and FGF2. MISCELLANEOUS: This protein binds heparin more strongly than does aFGF. SIMILARITY: Belongs to the heparin-binding growth factors family.

A high level of specificity for bFGF was shown by immunohistochemistry for this antiserum.
Application Information:
IHC (frozen), WB. Recommended to be used at a dilution of 1: 1000 to 1:2000 for both applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: rh Basic FGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge