Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-rh CNTF Sheep Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

S-025-100
BSENS-025-100EA 302 EUR InStock
BSENS-025-100
Anti-rh CNTF Sheep Polyclonal Antibody
Antikörper
CNTF is a survival promoting factor for different types of neurons in vitro and in vivo. The essential structural features for the biological function of human CNTF were investigated by Thier, M. et al. They showed that deletion of 14 N-terminal and 18 C-terminal amino acids significantly increased bioactivity compared to wild-type CNTF. FUNCTION: CNTF is a survival factor for various neuronal cell types. Seems to prevent the degeneration of motor axons after axotomy. SUBUNIT: Homodimer. SUBCELLULAR LOCATION: Cytoplasm. TISSUE SPECIFICITY: Nervous system. PHARMACEUTICAL: CNTF is being tested under the name Axokine by Regeneron Pharmaceuticals for treatment of human motor neuron diseases, such as amyotrophic lateral sclerosis (ALS). As it induces substantial weight loss, preferentially of fat as opposed to lean body mass, it is being used for obesity treatment. SIMILARITY: Belongs to the CNTF family.

This antibody specifically detects CNTF shown by western blot.
Application Information:
IHC, WB, ELISA. A dilution of 1:500 to 4000 is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: rh CNTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Sheep
Isotype: IgG
Reactivity: Rat, Mouse, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge