Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Ancient ubiquitous Protein 1 Sheep Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

S-035-100
BSENS-035-100EA 302 EUR InStock
BSENS-035-100
Anti-Ancient ubiquitous Protein 1 Sheep Polyclonal Antibody
Antikörper
AUP1 contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain thay may be involved in binding ubiquitin-conjugating enzymes.  The unprocessed precusor is of 476 amino acids in length and has an estimated molecular weight of 53 kDa. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing.TISSUE SPECIFICITY: Ubiquitous. SIMILARITY: Belongs to the AUP1 family. SIMILARITY: Contains 1 CUE domain.

This antiserum is specific for ancient ubiquitous protein 1.
Application Information:
IHC, WB. Use at a dilution of 1:500 to 1:6000. This antiserum works superbly in both paraffin embedded and frozen tissues. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Ancient ubiquitous Protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 48-60 of Human ancient ubiquitous protein 1
Host: Sheep
Isotype: IgG
Reactivity: Bovine, Sheep, Human, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge