Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-beta NGF Sheep Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

S-051-50
BSENS-051-50EA 312 EUR InStock
BSENS-051-50
Anti-beta NGF Sheep Polyclonal Antibody
Antikörper
FUNCTION: Nerve growth factor is important for the development and maintenance of the sympathetic and sensory nervous systems. It stimulates division and differentiation of sympathetic and embryonic sensory neurons. SUBUNIT: Homodimer, associated by noncovalent forces. SUBCELLULAR LOCATION: Secreted protein. SIMILARITY: Belongs to the NGF-beta family.

A cross reactivity of less than 1% to recombinant human BDNF, NT3, NT4/5 by ELISA has been shown.
Application Information:
IHC, 1-site ELISA, WB, immunoblot, inhibition of biological activity. A concentration of 1-3 µg/ml is recommended for IHC, western blot and immunoblot, ELISA, inhibition of biological activity in vitro. Use neat for in vivo studies at 2-10 µg/ml (ED50). This antiserum completely inhibits neuronal survival and the outgrowth actions of murine NGF in chicken DRG in vitro. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: beta NGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat, Human, Avis

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge