UST bench brush, soft, Vikan®
Lieferant: BASAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
45811 45812 45813 45814 45815 45816 45817 45818
BSNC45811EA 23.3 EUR InStock
BSNC45811 BSNC45812 BSNC45813 BSNC45814 BSNC45815 BSNC45816 BSNC45817 BSNC45818
UST bench brush, soft, Vikan®
Bürsten
This ULTRA SAFE TECHNOLOGY (UST) Bench Brush with Ultra Filament Security ensures effective sweeping of fine dry particles from conveyor belts, food preparation surfaces, tables and equipment in high-risk areas.
  • Soft bristles made from Polybutylene terephthalate (PBT)
  • Handle made from glass reinforced PP
  • Large choice of colours
  • Autoclavable at 121 °C
All UST brushes feature a unique bristle system that minimises the risk of contamination and bristle loss.
Zertifizierungen: Food contact approved. FDA compliant.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge