Contact slides, HYCON®, microbial monitoring of flat and curved surfaces in the food & beverage industry
Lieferant: BIOTEST
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.44023.0020 1.44018.0020 1.44019.0020 1.44022.0020 1.44021.0020 1.44023.0100 1.44022.0100
BTSM1.44023.0020EA 96.9 EUR InStock
BTSM1.44023.0020 BTSM1.44018.0020 BTSM1.44019.0020 BTSM1.44022.0020 BTSM1.44021.0020 BTSM1.44023.0100 BTSM1.44022.0100
Contact slides, HYCON®, microbial monitoring of flat and curved surfaces in the food & beverage industry
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
HYCON® Contact Slides for microbial monitoring of flat and curved surfaces are designed as standardised agar media in a flexible foil. Their unique design makes them universal tools for easy microbial monitoring of flat and curved surfaces. The choice of formulations includes total count media as well as selective media for yeasts and moulds, and for coliform organisms. Each contact slide is welded shrink-wrapped into a resealable sleeve to enable individual strips to be safely sterilised a number of times by peroxide gassing.
  • Individually sealed, H₂O₂-impermeable primary packaging
  • Resealable sleeve ensures sterility and enables safe handling and transportation
  • Produced in clean rooms, rigorously quality controlled, including 100% visual inspection of each contact slide
  • Growth promotion, packaging and storage extensively validated
  • Standardised contact area meets international guidelines (25 cm²)
  • Can be used for flat, curved or irregular surfaces
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge