Agar Strips, HYCON® for RCS air sampler
Lieferant: BIOTEST
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.44099.0025 1.44109.0040 1.44111.0100 1.44107.0050 1.44240.0050 1.44245.0025 1.44243.0050 1.44228.0040 1.44108.0040 1.44242.0050 1.44253.0050
BTSM1.44099.0025EA 0 EUR InStock
BTSM1.44099.0025 BTSM1.44109.0040 BTSM1.44111.0100 BTSM1.44107.0050 BTSM1.44240.0050 BTSM1.44245.0025 BTSM1.44243.0050 1.44228.0040 BTSM1.44108.0040 BTSM1.44242.0050 BTSM1.44253.0050
Agar Strips, HYCON® for RCS air sampler
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
A wide range of HYCON® Agar Strips are available specifically for RCS® Microbial Air Samplers. There are several total count media, selective media for yeasts and moulds, coliform organisms and staphylococci. Gamma-irradiated products in sterile double packaging are also available. Using standardised media which are manufactured in controlled cleanroom environments allows for the complete validation of the RCS® Systems according to ISO 14698-1/2. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments. HYCON® Agar Strips are used for monitoring of ambient air and compressed gases with the RCS® High Flow Touch Microbial Air Sampler and the RCS® Isolator. They are also applied with the RCS® Plus Ex Microbial Air Sampler for monitoring of explosion hazard areas.
  • Specifically designed for applications within isolator and clean room environments
  • Double packaging allows for easy transfer into critical areas
  • Contained additives for neutralisation of more than 5000 ppm of hydrogen peroxide (only TCI- γ)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge