Endotoxin standard, PyroDetect
Lieferant: BIOTEST
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.44161.0001
BTSM1.44161.0001EA 1020 EUR InStock
BTSM1.44161.0001
Endotoxin standard, PyroDetect
Standards Biologische Standards
PyroDetect is used to perform an endotoxin standard curve using different concentrations of endotoxin in dilution.
  • One of the two optional standards
  • Standard curve required to quantify the amount of pyrogens
  • Determine the limit of detection (LOD) of the PyroDetect System
To perfom the endotoxin standard curve, the PyroDetect Endotoxin Standard or any other International Reference Standard Endotoxin (RSE) can be used.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge