Petri dish platinum electrodes
Lieferant: Harvard Apparatus
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
45-2059 45-2060
BTXI45-2059EA 2810 EUR InStock
BTXI45-2059 BTXI45-2060
Petri dish platinum electrodes
Elektroden Elektroporationselektroden
These reusable oocyte Petri electrodes are ideal for electroporation of oocytes, embryos, or other cell types in small volumes.
  • Electrodes are permanently attached to a glass Petri dish and come with a glass lid. Designed to easily fit on an inverted microscope stage to allow easy observation of cells during electroporation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge