Wash bottles, wide neck, with printed name, LaboPlast®
Lieferant: BURKLE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0310-3050 0310-3054 0310-3053 0310-3052 0310-3055 0310-3051
BURK0310-3050EA 9.45 EUR InStock
BURK0310-3050 BURK0310-3054 BURK0310-3053 BURK0310-3052 BURK0310-3055 BURK0310-3051
Wash bottles, wide neck, with printed name, LaboPlast®
Flaschen Spritzflaschen
LDPE yellow transparent with light protection.
  • Safe filling due to wide-necked opening
  • Good chemical resistance
  • Good temperature resistance
Wash bottle with medium overprint as well as hazard and safety instructions (health statements/physical statements) based on the GHS system with substance name in German, English, French and Spanish, GHS pictograms and CAS number.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge