Measuring spoons, stainless steel
Lieferant: BURKLE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5334-8000
BURK5334-8000EA 15.95 EUR InStock
BURK5334-8000
Measuring spoons, stainless steel
Löffel
Stainless steel V2A. Set of four measuring spoons. Ideal for fast and easy measuring from 1,25 to 15 ml.
  • High-gloss finished, handle matt polished
  • Volume is engraved on the handle of each measuring spoon
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge