Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Tubing couplings, rapid, NW 3,2 mm

Lieferant: BURKLE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

8741-1204 8751-1104 8751-1101 8741-1202 8741-1002 8751-1102 8751-1002 8742-1104 8752-1004 8741-2002 8751-2102 8751-2104 8743-2004 8740-1122 8740-1141 8740-1142 8755-1104 8751-1001 8751-1004 8751-1003 8751-1103 8751-2101 8753-1102 8753-1103 8753-1104 8740-2022 8752-2104 8752-1142 8743-1101 8744-1004 8752-1104 8741-1203 8743-1103 8755-1102 8743-1104 8741-1004 8743-1102 8743-2001 8750-1022 8742-2004 8741-1003
BURK8741-1204EA 18.05 EUR InStock
BURK8741-1204 BURK8751-1104 BURK8751-1101 223-1126 223-1127 223-1128 223-1129 BURK8742-1104 BURK8752-1004 BURK8741-2002 BURK8751-2102 BURK8751-2104 BURK8743-2004 BURK8740-1122 BURK8740-1141 BURK8740-1142 BURK8755-1104 BURK8751-1001 BURK8751-1004 BURK8751-1003 BURK8751-1103 BURK8751-2101 BURK8753-1102 BURK8753-1103 BURK8753-1104 BURK8740-2022 BURK8752-2104 BURK8752-1142 BURK8743-1101 BURK8744-1004 BURK8752-1104 BURK8741-1203 BURK8743-1103 BURK8755-1102 BURK8743-1104 BURK8741-1004 BURK8743-1102 BURK8743-2001 BURK8750-1022 BURK8742-2004 BURK8741-1003
Tubing couplings, rapid, NW 3,2 mm
Fittings und Verbinder
Spring/lock Stainless steel V4A

Fast and safe connection for liquids, gases, compressed air and vacuum. Rapid couplings are the ideal when connecting and separating hoses, machines, apparatus, containers, etc. Top quality, with tested reliability, small space requirement, maximum possible flow, easy operation (single handed operation) and an elegant design distinguish the rapid couplings.The couplings are available in two different materials: POM (Polyoxymethylene) and PP (Polypropylene). The rapid couplings are available with a hose nozzle, hose screw joint, thread connector or as a built-in component for control panels, with or without stop-valve respectively.The couplings are simply connected to each other, automatically clicking intoplace and opening the valves.When disconnecting, (one handed operation), the valves are automatically sealed, no leakage occurs.

POM
Seal (standard): Buna-N
Operating pressure at 20 °C with vacuum up to 10 bar
Operating temperature: -40 °C to +80 °C

PP
Seal (standard): EPDM
Operating pressure at 20 °C: up to 8 bar
Operating temperature: 0 °C to +110 °C

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge