Measuring scoops
Lieferant: BURKLE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9614-0005 9614-0050 9614-0010 9614-0500 9614-0025 9614-0100 9614-0250 9614-1000
BURK9614-0005EA 0.59 EUR InStock
BURK9614-0005 BURK9614-0050 BURK9614-0010 BURK9614-0500 BURK9614-0025 BURK9614-0100 BURK9614-0250 BURK9614-1000
Measuring scoops
Schaufeln
Universal graduated PP shovels for filling and measuring of powders and granules. For exact metering, the graduated shovel is to be overfilled and the excess at the upper edge of the shovel is to be wiped off.
  • High chemical resistance
  • Calibrated volume when full to the brim
  • Suitable for food contact
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge