Anti-DnaJB4 Rabbit Polyclonal Antibody [clone: R120]
Lieferant: Bioworld Technology
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS3025
BWRLBS3025EA 262 EUR InStock
BWRLBS3025
Anti-DnaJB4 Rabbit Polyclonal Antibody [clone: R120]
Antikörper
Synthetic peptide, corresponding to amino acids 90-132 of Human DnaJB4.
DnaJB4 (R120) pAb detects endogenous levels of DnaJB4 protein.
Type: Primary
Antigen: DnaJB4
Clonality: Polyclonal
Clone: R120
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge