Anti-beta-1,4-Gal-T3 Rabbit Polyclonal Antibody [clone: F304]
Lieferant: Bioworld Technology
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS3090
BWRLBS3090EA 262 EUR InStock
BWRLBS3090
Anti-beta-1,4-Gal-T3 Rabbit Polyclonal Antibody [clone: F304]
Antikörper
Synthetic peptide, corresponding to amino acids 281-320 of Human beta-1,4-Gal-T3.
?-1,4-Gal-T3 (F304) pAb detects endogenous levels of?-1,4-Gal-T3 protein.
Type: Primary
Antigen: beta-1,4-Gal-T3
Clonality: Polyclonal
Clone: F304
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge