Citalopram-D6 hydrobromide solution, certified reference material, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-090-1ML
C-090-1MLEA 219 EUR InStock
C-090-1ML
Citalopram-D6 hydrobromide solution, certified reference material, Supelco®
Standards Organische Standards
Citalopram is an antidepressant sold under the trade names Celexa® and Cipramil for the treatment of major depression.
Citalopram is a selective serotonin reuptake inhibitor, a class of drugs that also includes fluoxetine, paroxetine, and sertraline. This internal standard is suitable for quantitation of citalopram levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
Verpackung: Snap-N-Spike®/Snap-N-Shoot®
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=280ee1c3fa6facd8fca10e966c8fd83fb73a466f&vid=280ee1c3fa6facd8fca10e966c8fd83fb73a466f&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge