Human pulmonary artery endothelial cells (HPAEC)
Lieferant: PROMOCELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-14024
C-14024EA 380 EUR InStock
C-14024
Human pulmonary artery endothelial cells (HPAEC)
Zellen
Primary Human Pulmonary Artery Endothelial Cells (HPAEC) are isolated from human pulmonary arteries (the main pulmonary artery, left and right branches)
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
Since the pulmonary arteries carry blood from the heart to the lungs, they are the only arteries (other than umbilical arteries) that carry deoxygenated blood.
Bestellinformation: Human Pulmonary Artery Endothelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge