Human aortic adventitial fibroblasts (HAoAF)
Lieferant: PROMOCELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-14037
C-14037EA 332 EUR InStock
C-14037
Human aortic adventitial fibroblasts (HAoAF)
Zellen
Primary Human Aortic Adventitial Fibroblasts (HAoAF) are isolated from human aortic adventitial tissue. HAoAF can be used for studies concerning vascular diseases e.g. neointimal formation, vascular remodeling, atherosclerosis, and hypertension. They are also often used for the examination of fibroblast disorders like fibrosis or other diseases linked to either imperfect or excessive accumulation of fibroblasts.
Applications include: Wound healing research, tissue engineering research, regeneration studies, toxicological research, oncology, epithelial-mesenchymal interaction studies.
Bestellinformation: Human Aortic Adventitial Fibroblasts can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge