Human coronary artery smooth muscle cells
Lieferant: PROMOCELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-14052
C-14052EA 370 EUR InStock
C-14052
Human coronary artery smooth muscle cells
Zellen
Primary human coronary artery smooth muscle cells (HCASMC) are isolated from human coronary arteries and stain positive for smooth muscle α-actin. Coronary arteries supply the heart with blood and obstruction in atherosclerosis causes myocardial infarction. HCASMC are qualified to study the function of smooth muscle cells in the development of atherosclerotic plaques or in other diseases, like hypertension.
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
Pellets are stored in RNAlater®, an aqueous protection solution that rapidly permeates the cells and stabilises cellular DNA, RNA and proteins and protects them against degradation. Pellets are useful for a wide range of research applications, such as gene expression analysis, cell characterisation studies and protein characterisation experiments.
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
They are also useful for stent-graft compatibility testing.
Bestellinformation: Human Coronary Artery Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge