Skeletal muscle cells (SkMC), human
Lieferant: PROMOCELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-14060
C-14060EA 341 EUR InStock
C-14060
Skeletal muscle cells (SkMC), human
Zellen
Primary human skeletal muscle cells (SkMC) are isolated from different skeletal muscles (eg. musculus pectoralis major) from adult single donors. They are positive for sarcomeric myosin and negative for smooth muscle specific α-actin.
Primary human skeletal muscle cells (SkMC) are optimal for in vitro muscle studies. They proliferate very well in the mitogen-rich skeletal muscle cell growth medium. Fusion to myotubes with typical multinucleated syncytia can be induced by using the skeletal muscle cell differentiation medium.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge