Human renal epithelial cells (HREpC)
Lieferant: PROMOCELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-14067
C-14067EA 324 EUR InStock
C-14067
Human renal epithelial cells (HREpC)
Zellen
Primary human renal epithelial cells (HREpC) are isolated from the adult kidney and stain positive for cytokeratin.
  • Cytokeratin positive
  • Routinely morphology checked
  • Cryopreserved at second passage
Applications: Drug uptake studies, toxicological research, wound healing research, oncology, cell differentiation studies, respiratory disease research
Kidneys have a very complex structure and contain diverse epithelia such as those in the convoluted tubules, the cortical collecting ducts, the calyxes, and the renal pelvis. The epithelial cells present in the distinct ducts and tubules differ in their morphology and physio-logical function. Consequently, HREpC are comprised of a heterogeneous population of epithelial cells. Important physiological processes like osmoregulation and excretion can be studied with HREpC in vitro.
Bestellinformation: Human renal epithelial cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge