Human chondrocytes
Lieferant: PROMOCELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-14070
C-14070EA 367 EUR InStock
C-14070
Human chondrocytes
Zellen
Primary human chondrocytes (HCH) are isolated from normal human articular cartilage from the knee and hip joints. HCH are routinely tested for the capacity to differentiate in 3D spheroids.
The articular cartilage covers the joints between bones and contains no blood vessels, lymphatic vessels, or nerve fibers. It is composed of only one specialised cell type, the chondrocytes, which produce and maintain the extracellular matrix of cartilage, i.e. collagen (mostly type II) and proteoglycans (primarily aggrecan).
Applications include: Osteoarthritis research, joint research, physiology studies, chondrocyte differentiation experiments, hyaline cartilage investigations.
Lieferumfang: Human Chondrocytes can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge