Human mesenchymal stem cells from adipose tissue
Lieferant: PROMOCELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-14092
C-14092EA 361 EUR InStock
C-14092
Human mesenchymal stem cells from adipose tissue
Zellen
Human Mesenchymal Stem Cells from Adipose Tissue (hMSC-AT) are harvested from normal human adipose tissue. The cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes and osteoblasts.
Applications include: Stem cell research, regenerative medicine research, tissue engineering studies, developmental experiments.
PromoCell's hMSC-BM cells show a verified marker expression profile that complies with ISCT* recommendations, providing well characterised cells.
*ISCT (International Society for Cellular Therapy) Cytotherapy (2006) Vol. 8, No. 4, 315-317
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge