Human mononuclear cells from cord blood (hMNC-CB)
Lieferant: PROMOCELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-14096
C-14096EA 239 EUR InStock
C-14096
Human mononuclear cells from cord blood (hMNC-CB)
Zellen
Human mononuclear cells from cord blood (hMNC-CB) are isolated from umbilical cord blood, which has a much higher content of primitive progenitor cells compared to adult peripheral blood.
Applications include: Stem cell research, transplantation basic research, hematologic and immunologic studies, oncology research, virology and infection biology.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge