Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Direct Blue 1, Pulver

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

C8679-25G
C8679-25GEA 194 EUR InStock
C8679-25G
Direct Blue 1, Pulver
Direct blue 1
Chicago Sky Blue 6B is a counterstain for background autofluorescence in fluorescence and immunofluorescence histochemistry. It has also been used as a filling in glass pipette electrodes for electrophysiological recordings in the brain sections of rats.

Chicago Sky Blue 6B is a large organic acid, structurally related to glutamate, that is a potent and efficient competitive inhibitor of vesicular glutamate uptake.


Formel: C₃₄H₂₈N₆O₁₆S₄
MW: 992.79 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00004020
CAS-Nummer: 2610-05-1
Order Now
Spezifikation Testergebnisse
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Colour)
10 mg/ml in Water
Dark Blue
Carbon 32.7 - 49.6%
Nitrogen 6.5 - 10.2%
Dye Content Approximately 80%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge