Cooling bags, Fold’N Cool™
Lieferant: CAMPING GAZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2000011722 2000011725 2000011723 2000011724
CAMP2000011722EA 18 EUR InStock
CAMP2000011722 CAMP2000011725 CAMP2000011723 CAMP2000011724
Cooling bags, Fold’N Cool™
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
The Fold'N Cool™ soft foldable coolers are comfortable to carry and are available in various sizes and could be folded to store after use.
  • Adjustable shoulder strap
  • Collapsible cooler
  • EPE closed cells foam 6 mm
The bags with 10 litre capacity are light, which holds 8×0,5 litre bottles vertically and 10×0,5 litre bottles horizontally. The bags with 30 litre capacity can hold 9×1,5 litre bottles vertically and 12×1,5 litre bottles horizontally. The bags with 20 litre capacity can hold 6×1,5 litre bottles vertically and 5×1,5 litre bottles horizontally. The bags with 5 litre capacity can hold 6×0,33 litre bottles vertically and horizontally.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge