Bunsen burners, LABOGAZ 206
Lieferant: CAMPING GAZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2000020656
CAMP2000020656EA 84 EUR InStock
CAMP2000020656
Bunsen burners, LABOGAZ 206
Heizgeräte Gasbrenner
For use with Campingaz® pierceable cartridge C 206 GLS. Ideal use in wide range of areas such as laboratories, schools, jewelers, opticians and handicrafts.
  • Attached with clip and go installation system which makes changing cartridges easy
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge