Bunsenbrenner, Labogaz® 470
Lieferant: CAMPING GAZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2000020657
CAMP2000020657EA 110 EUR InStock
CAMP2000020657
Bunsenbrenner, Labogaz® 470
Heizgeräte Gasbrenner
Compact bunsen burner for operation with the Campingaz cartridge CV 470 Plus.
  • Device can be removed from the cartridge even if the cartridge is not yet completely empty
  • Ideal for heating solutions in glass flasks and test tubes or bending parts such as glass tubes
For a wide range of applications in crafts (optics, goldsmith work), industrial laboratories (physics, chemistry, pharmaceuticals) or schools (practical work, experiments).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge