Cooling box, electrical, Powerbox® Plus
Lieferant: CAMPING GAZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2000037453 2000037452
CAMP2000037453EA 121 EUR InStock
CAMP2000037453 CAMP2000037452
Cooling box, electrical, Powerbox® Plus
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
Electrical cooling box equipped with a dial wheel, with which the power and efficiency of the box can be regulated helps to keeps the contents fresh and protected from odour, mold and fungi.
  • Premium PU insulated lid for even better performance
  • It can be used both on 12 V (auto) and 230 V (normal) socket outlet
  • Suitable for longer camping trips or outings
  • Bail handle for comfortable carrying
Antimicrobial effect is obtained from an additive which is melted down into the plastic before it is molded into the liner.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge