Self Seal Sterilisation Pouches
Lieferant: CARDINAL HEALTH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92308
CARD92308EA 72 EUR InStock
CARD92308
Self Seal Sterilisation Pouches
Beutel Sterilisationsbeutel
These sterilization pouch combines a blue film and a medical grade paper to provide breathability, sterile barrier and indication of processing. To open, a convenient thumb notch is positioned at the top of the pouch, above the chevron seal. Each pouch includes an external process indicator for steam and ethylene oxide (EO) gas sterilisation.
  • Pouched sealed mechanically or manually with a heat sealer
  • For use in steam and gas (EO) sterilisers
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge