10009292-96S
10009292-96
10009292-480S
10009292-480
CAYM10009292-96SEA
454
EUR
InStock
CAYM10009292-96S
CAYM10009292-96
CAYM10009292-480S
CAYM10009292-480
Leukotriene B₄ Express ELISA kit
Tests
ELISAs
A competitive assay that can be used for quantification of LTB₄ in plasma and other sample matrices.
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure LTB₄ levels down to 45 pg/ml
- Rapid assay; get results in under five hours
LTB₄ stimulates a number of leukocyte functions, including aggregation, stimulation of ion fluxes, enhancement of lysosomal enzyme release, superoxide anion production, chemotaxis, and chemokinesis.
LTB₄ is metabolized in leukocytes and hepatocytes to less active 20-hydroxy and 20-carboxy LTB₄ and then further oxidized to ω-carboxy dinor LTB₄ and ω-carboxy tetranor-LTB3. It is not excreted in the urine.
Lieferumfang: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, ellman's reagent, tracer and antiserum dye.