Leukotriene B₄ Express ELISA kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10009292-96S 10009292-96 10009292-480S 10009292-480
CAYM10009292-96SEA 454 EUR InStock
CAYM10009292-96S CAYM10009292-96 CAYM10009292-480S CAYM10009292-480
Leukotriene B₄ Express ELISA kit
Tests ELISAs
A competitive assay that can be used for quantification of LTB₄ in plasma and other sample matrices.
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure LTB₄ levels down to 45 pg/ml
  • Rapid assay; get results in under five hours
LTB₄ stimulates a number of leukocyte functions, including aggregation, stimulation of ion fluxes, enhancement of lysosomal enzyme release, superoxide anion production, chemotaxis, and chemokinesis.
LTB₄ is metabolized in leukocytes and hepatocytes to less active 20-hydroxy and 20-carboxy LTB₄ and then further oxidized to ω-carboxy dinor LTB₄ and ω-carboxy tetranor-LTB3. It is not excreted in the urine.
Lieferumfang: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, ellman's reagent, tracer and antiserum dye.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge